Learn More
OHAUS™ Temperature Probe
Hot plates and stirrers accessory
Brand:  OHAUS™ 30500593
3860.60 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16620482
  • 3965.00 NOK / Each
In Stock
Explore more special offers
Description
  • Temperature probe in stainless steel or PTFE
  • Varying sizes available
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 cm
Temperature Probe
PTFE
Product Title
OHAUS™ Temperature Probe > 25 cm, PTFE