Learn More
OHAUS™ Temperature Probe
Hot plates and stirrers accessory
Brand:  OHAUS™ 30500591
3041.68 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16600482
  • 3125.00 NOK / Each
In Stock
Explore more special offers
Description
  • Temperature probe in stainless steel or PTFE
  • Varying sizes available
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 cm
Temperature Probe
PTFE
Documents
One moment while we fetch your results.
Product Title
OHAUS™ Temperature Probe > 20 cm, PTFE