missing translation for 'onlineSavingsMsg'
Learn More
TEMED, Approx. 99%, Molecular Biology Reagent, MP Biomedicals™ Product Code.: 11406501

TEMED, Approx. 99%, Molecular Biology Reagent, MP Biomedicals™

Product Code. 11406501 Shop All MP Biomedicals Products
100 mL, 100mL
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 11406501

Brand: MP Biomedicals 0219401980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N,N,N',N'-Tetramethylethylenediamine is used to catalyze the formation of free radicals from ammonium persulfate or riboflavin

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Density 0.775g/mL at 20°C
Assay Percent Range Approx. 99%
Quantity 100 mL
Formula Weight 116.2
Grade Molecular Biology Reagent
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine, FTIR
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TEMED, Approx. 99%, Molecular Biology Reagent, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.