Learn More
Kimberly-Clark™ Tecnol™ Fluidshield™ Pleat-style Facemask with Earloops
Non-sterile pleat-style masks with excellent protection against blood or fluid splash. The surgical masks are EN14683 Type IIR certified and feature breathable LONCET™ material, providing excellent fluid resistance and a clear Splashguard™ visor.
Specifications
Color | Orange |
---|---|
Sterility | Non-sterile |
Certifications/Compliance | ISO 14001, ISO 9001; Industrial Use Only |
Includes | Splashguard™ visor |
Latex Free | Yes |
Description
- Splashguard™ visor attached to the Fluidshield™ mask shields the eyes from blood or fluid splash.
- Excellent particle and bacterial filtration efficiency to ensure the wearer’s security.
- Certified according to EN14683 Type IIR, and provided with supporting technical information to demonstrate regulatory compliance.
- Constructed from a low-lint polyethylene film outer layer for enhanced protection.
- Contain no natural rubber latex or silicone, reducing the risks of skin irritation for the wearer.
- The masks are provided in a single universal size.
Specifications
Orange | |
ISO 14001, ISO 9001; Industrial Use Only | |
Yes |
Non-sterile | |
Splashguard™ visor |
Product Title
Kimberly-Clark™ Tecnol™ Fluidshield™ Pleat-style Facemask with Earloops