missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Taurine
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Brand: Melford T38200-1000.0
This item is not returnable.
View return policy
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- Substance Name: 2-Aminoethanesulfonic acid
- CAS Number: 107-35-7
- Molecular Formula: C2H7NO3S
- Molecular Weight: 125.15 g/mol
Specifications
107-35-7 | |
C2H7NO3S | |
1 kg |