Learn More
Applied Biosystems™ TaqMan™ Gene Expression Control Kit
The Applied Biosystems™ TaqMan™ Gene Expression Control Kit is designed to detect and quantify eukaryotic 18S ribosomal RNA (rRNA) and human telomerase reverse transcriptase (hTERT) in human Raji cDNA on the QuantStudio™ 5 Real-Time PCR System.
Brand: Applied Biosystems™ A29349
2021.25 NOK valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Gene-specific, ready-to-use TaqMan gene expression assays provide you with a fast, simple way to measure changes in gene expression in real-time PCR experiments.
Features of the TaqMan Gene Expression Control Kit include:
• Simple—includes pre-formulated assays with our universal cycling conditions
• Easy to use—just add to your real-time PCR experiment with no additional steps
• Robust—generate reproducible quantitative gene expression data
Ready-to-use gene expression solution
The TaqMan Gene Expression Control Kit contains TaqMan gene expression assays for eukaryotic 18S rRNA and hTERT and a sample of human Raji cDNA for use in TaqMan-based relative quantitation on the QuantStudio 5 Real-Time PCR System.
Specifications
Store in freezer (-5°C to -30°C). | |
Gene Expression Control Kit | |
TaqMan™ | |
Human | |
Gene Expression |
Tube | |
FAM | |
Approved for shipment on Wet or Dry Ice | |
18 S rRNA & hTERT | |
250 Reactions |