Learn More
Tanshinone IIA sulfonate sodium, MedChemExpress
MedChemExpress Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA, which acts as an inhibitor of store-operated Ca2+ entry (SOCE), and is used to treat cardiovascular disorders.
Brand: Medchem Express HY-N1370
This item is not returnable.
View return policy
Description
Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA, which acts as an inhibitor of store-operated Ca2+ entry (SOCE), and is used to treat cardiovascular disorders.Specifications
Tanshinone IIA sulfonate sodium | |
wine | |
C19H17NaO6S | |
Sodium Tanshinone IIA sulfonate Tanshinone IIA sodium sulfonate | |
O=S(C1=C(C)C(C(C(C2=C3C=CC4=C2CCCC4(C)C)=O)=O)=C3O1)(O[Na])=O | |
396.39 | |
Research | |
COVID-19-immunoregulation |
69659-80-9 | |
Research | |
10 mg | |
DMSO : 62.5 mg/mL (157.67 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (21.01 mM; Need ultrasonic) | |
396.39 | |
98.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.