Learn More
Tanshinone I, MedChemExpress
MedChemExpress Tanshinone I is an inhibitor of type IIA human recombinant sPLA2 (IC50=11 μM) and rabbit recombinant cPLA2 (IC50=82 μM).
Brand: Medchem Express HY-N0134
This item is not returnable.
View return policy
Description
Tanshinone I is an inhibitor of type IIA human recombinant sPLA2 (IC50=11 μM) and rabbit recombinant cPLA2 (IC50=82 μM).Specifications
Tanshinone I | |
Dark Brown | |
C18H12O3 | |
Tanshinone A | |
O=C(C1=C2C=CC3=C1C=CC=C3C)C(C4=C2OC=C4C)=O | |
276.29 | |
Research | |
COVID-19-immunoregulation |
568-73-0 | |
Research | |
5 mg | |
DMSO : 2 mg/mL (7.24 mM; Need ultrasonic) | |
276.29 | |
98.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.