Learn More
DWK Life Sciences Tamper Evident Screw Cap, Narrow Mouth, Red PP, Vented Closure, for Soda-Lime Glass Screw-Top Bottles
Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 32
GL 45
Description
Vented red polypropylene screw cap for narrow mouth/high form DIN thread soda lime glass bottles. PTFE membrane for two way gas flow. Closed-cell polyolefin foam cap liner for effective sealing with bottle neck.
- Vented with PTFE membrane to ensure safety with two way gas transfer capability
- Closed-cell polyolefin foam cap liner for effective seal
- Tamper evident ring
- High form DIN thread
Specifications
Specifications
For Use With (Application) | Storage of samples and reagents; Sample collection; Transport and shipping |
Material | Polypropylene |
Quantity | 10/Pk. |
Thread Size (Metric) | GL 32 |
Autoclavable | Autoclavable |
Diameter (Metric) | 45 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
DWK Life Sciences Tamper Evident Screw Cap, Narrow Mouth, Red PP, Vented Closure, for Soda-Lime Glass Screw-Top Bottles >