Description
The RPB T200 enables you to work in your environment free of respiratory hazards and discomfort through its supportive design.
- Very lightweight and comfortable to help protect the user better. It provides a wide viewing area to make your work easier.
- Multi adjustment to fit a wide range of users.
- Certified to EN 12941 1998 + A2:2008 (TH3) and EN 166 2001 1 F 3
- Includes: Tychem 2000 face seal, head harness assembly, PX5™ breathing tube
- To use with PAPR PX5™
Specifications
Specifications
Color | White |
Product Type | PAPR System with Hood and Head Harness |
Size | One Size |
Product Content Correction
Product Title
GVS T200 Respirator and Head Harness >