Learn More
GVS T200 Respirator with Full Hood and Internal Face Seal
Click to view available options
:
GVS T200 Respirator
Description
The T200 respirator provides great protection for those in healthcare, EMS, and other types of medical enviroments.
- Very lightweight and comfortable to help protect the user better. It provides a wide viewing area to make your work easier.
- Multi adjustment to fit a wide range of users.
- Certified to EN 12941 1998 +A2:2008 and EN 166 2001 1 F 3.
- Includes: Full hood with internal face seal, air duct and head harness assembly, 04-831 PX5™ breathing tube
Specifications
Specifications
Color | White |
Product Type | PAPR System with Hood and Head Harness |
Size | One Size |
Product Content Correction
Product Title
GVS T200 Respirator with Full Hood and Internal Face Seal >