Learn More
Lonza™ SYPRO™ Protein Gel Stains
Liquid format protein gel stains
Brand:  Lonza™ 50543
 View more versions of this product
Product Code. 11500777
  • 6425.00 NOK / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Description
SYPRO Ruby Protein Gel Stains
  • Ideal for 2-D gel analysis Sensitivity rivals silver stain Detects nanogram levels of protein Offers a broad linear quantitation range and consistent gel-to-gel staining for accurate protein expression comparisons
SYPRO Tangerine Protein Gel Stain
  • Provides high sensitivity for detection of proteins in SDS-polyacrylamide gels
  • Does not require the use of acids or organic solvents
  • Suitable for use with saline solutions Ideal for use when eluting active protein complexes from gels because the mild staining conditions are unlikely to affect protein activity
SYPRO Red Protein Gel Stain
  • Detects as little as 4 to 8ng of protein per band or proteins as small as 6.5kDa Provides linear protein quantitation up to three orders of magnitude, from ng to μg per band
  • Offers complete staining in less than one hour—no fixation or destaining required
  • Compatible with many types of detection systems Recommended for instruments that employ frequency-doubled Nd:YAG or green or red He-Ne lasers
Product licensed by Cambrex from Molecular Probes, Inc. For research and quality control use only.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red
Liquid
500 μL
Protein Stain
5000X Concentrated stock solutions in DMSO
4°C