missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14988-1-AP-20UL
This item is not returnable.
View return policy
Description
STX4 is a plasma membrane t-SNARE that mediates docking of transport vesicles.STX4 is necessary for the translocation of SLC2A4 from intracellular vesicles to the plasma membrane. Together with STXB3 and VAMP2, STX4 may also play a role in docking/fusion of intracellular GLUT4-containing vesicles with the cell surface in adipocytes. STX4 may also play a role in docking of synaptic vesicles at presynaptic active zones.Specifications
Syntaxin 4 | |
Polyclonal | |
Unconjugated | |
STX4 | |
p35 2, STX4, STX4A, syntaxin 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20909, 6810, 81803 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70452, Q08850, Q12846 | |
STX4 | |
Syntaxin 4 Fusion Protein Ag6973 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction