missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16013-1-AP-150UL
This item is not returnable.
View return policy
Description
Syntaxins, a family of proteins involved in the fusion of synaptic vesicles with the plasma membrane, display broad tissue distribution and contain carboxy-terminal hydrophobic domains that direct themselves to their respective intracellular compartments. Syntaxin 18 mainly localizes to the endoplasmic reticulum (ER), and functions in transport between the ER and Golgi. Syntaxin 18 also plays a role in ER-mediated phagocytosis, possibly by regulating the specific fusion of the ER and plasma or phagosomal membranes.Specifications
Syntaxin 18 | |
Polyclonal | |
Unconjugated | |
STX18 | |
STX18, syntaxin 18, Ufe1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360953, 53407, 71116 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68FW4, Q8VDS8, Q9P2W9 | |
Stx18 | |
Syntaxin 18 Fusion Protein Ag8777 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction