Learn More
KARTELL Sylicon Conical Stopper
The Sylicon Conical Stopper creates a seal in bottles and flasks.
Brand:  KARTELL 0380500
 View more versions of this product
Product Code. 10737482
  • 202.00 NOK / Pack of 10
Only null left
Estimated Shipment: 14-08-2025
Description
  • Autoclavable.
  • SUITABLE FOR FOODSTUFF.
  • MATERIAL: SYLICON
Specifications
Specifications
Autoclavable
13 mm
18 mm
Non-sterile
White
18 mm
Silicone
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
KARTELL Sylicon Conical Stopper > Top Diameter: 18 mm