Learn More
KARTELL Sylicon Conical Stopper
The Sylicon Conical Stopper creates a seal in bottles and flasks.
Brand:  KARTELL 0380700
 View more versions of this product
Product Code. 10727672
  • 438.00 NOK / Pack of 10
Only null left
Estimated Shipment: 13-08-2025
Description
  • Autoclavable.
  • SUITABLE FOR FOODSTUFF.
  • MATERIAL: SYLICON
Specifications
Specifications
Autoclavable
16 mm
23 mm
Non-sterile
White
23 mm
Silicone
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
KARTELL Sylicon Conical Stopper > Top Diameter: 23 mm