missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Swine Diet, Refined, Pellet

Swine diet. Contains casein, lard, special salt mix and special MP vitamin fortification.

11710.00 NOK - 44060.00 NOK

Specifications

Product Type Animal Diet
Autoclavable Autoclavable
Disposable Single-use
Format Pellet
Sterility Non-sterile
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11467221
View Documents
MP Biomedicals
0296028410
10 kg
11710.00 NOK
10kg
Please to purchase this item. Need a web account? Register with us today!
11477221
View Documents
MP Biomedicals
0296028420
20 kg
20245.00 NOK
20kg
Please to purchase this item. Need a web account? Register with us today!
11487221
View Documents
MP Biomedicals
0296028450
50 kg
44060.00 NOK
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Swine diet
  • Contains casein, lard, special salt mix and special MP Bio vitamin fortification
Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Autoclavable
Pellet
Room Temperature
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.