missing translation for 'onlineSavingsMsg'
Learn More

SURF5 Mouse anti-Human, Clone: PCRP-MED22-1E4, Novus Biologicals™

Mouse Monoclonal Antibody

2355.00 NOK - 4680.00 NOK

Specifications

Antigen SURF5
Clone PCRP-MED22-1E4
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 0.5-2 ug/million cells, ELISA, Immunofluorescence 0.5-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18329606
View Documents
Bio-Techne
NBP3-13787-20UG
20 μg
2355.00 NOK
20µg
Please to purchase this item. Need a web account? Register with us today!
18320883
View Documents
Bio-Techne
NBP3-13787-100UG
100 μg
4680.00 NOK
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SURF5 Monoclonal antibody specifically detects SURF5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein Array
Specifications

Specifications

SURF5
0.2 mg/ml
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array
Unconjugated
Mouse
Human
Q15528
6837
IgG2c
Protein A or G purified
PCRP-MED22-1E4
Western Blot 1-2 ug/ml, Flow Cytometry 0.5-2 ug/million cells, ELISA, Immunofluorescence 0.5-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
Med24, mediator complex subunit 22SURF5surfeit 5, mediator of RNA polymerase II transcription subunit 22, MGC48682, Surf-5, Surfeit locus protein 5
Recombinant full-length human SURF5 protein (Uniprot: Q15528)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.