Learn More
Fisherbrand™ SureOne™ Filter Tip Reload Pipette Tips
New, improved Fisherbrand SureOne Reload system offers a full line of micropoint universal fit pipet tips
392.00 NOK - 670.00 NOK
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Color | Clear |
Autoclavable | Autoclavable |
Format | Refill |
Product Code | Brand | Volume (Metric) | Tip Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Tip Style | Price | Quantity & Availability | ||||
11927724
|
Fisherbrand™
|
0.1 to 20 μL | Extended Length |
670.00 NOK
Pack of 960 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
11967724
|
Fisherbrand™
|
30 to 300 μL | Extended Length |
670.00 NOK
Pack of 960 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
11917724
|
Fisherbrand™
|
0.1 to 10 μL | Extended Length |
534.00 NOK
Pack of 960 |
In Stock
Log in to see stock. |
||||
11977724
|
Fisherbrand™
|
100 to 1000 μL | Extended Length |
392.00 NOK
Pack of 560 |
In Stock
Log in to see stock. |
||||
Description
Sterile and free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin and DNA
Packaging Features
- Offset wells allow filter tips to stack
- Rigid inserts are multi-channel format ensuring secure tip seal
- Lot numbered inserts for traceability also include easy to read tip volume and Fisher catalog number
- Sterile packaging with outer sleeve protects tips from contamination while refilling inserts
- Environmentally friendly reload packaging reduces plastic waste and saves space
- Blister sleeve guide rails align tip inserts for smooth and reliable transfer into the rack
- Flush Fit Inserts sit flush with rack, allowing easy multichannel pipetting
- Recycle Logo molded into all rack components
- Notched clasp props lid open for autoclave venting
- Sure Stack lid rim prevents racks from sliding around when stacked and carted around a lab
- Safe-Lock latch allows for one-handed open and closing ability and won't pop open when dropped
- Sure-snap design securely holds insert in place, preventing it from being lifted out while pipetting
- Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
- Inserts are easy to place onto and remove from rack; clip onto rack for a secure fit
- Lot traceable—all inserts contain the Fisherbrand part number, tip volume, lot number
- Indented insert holes inhibit tip movement, stabilizing them for multichannel work
- Contact Clip helps prevent unnecessary contact with the tips while clipping in inserts
- Multichannel design; all inserts are 96-well format
- Robust non-pliable design provides secure seals on every tip
Specifications
Filtered | |
Clear | |
Refill | |
Yes | |
Polypropylene |
Sterile | |
Autoclavable | |
Extended Length | |
SureOne |