Learn More
Fisherbrand™ Mixing Cup Head
Facilitates quick and safe mixing. For use with the Wizard™ Infrared Vortex Mixers.
Brand:  Fisherbrand™ 10002021
139.96 NOK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Product Code. 11726105
  • 160.16 NOK / Each
Only null left
Estimated Shipment: 07-07-2025
Add to basket
Description
Description
  • Tube containing sample placed on cup head
  • Can be easily substituted by any of the different accessories available for mixing tubes of different sizes
  • For use with Vortex Mixers
Specifications
Specifications
Mixing Cup Head
Plastic
50 mm
CLASSIC and WIZARD vortex mixers
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Mixing Cup Head > Diameter: 50 mm