missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT1C3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
4250.00 NOK
Specifications
Antigen | SULT1C3 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SULT1C3 Polyclonal specifically detects SULT1C3 in Human samples. It is validated for Western Blot.Specifications
SULT1C3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
EC 2.8.2, EC 2.8.2.1, EC 2.8.2.2, ST1C3, Sulfotransferase 1C3, Sulfotransferase Family, Cytosolic, 1C, Member 3 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human SULT1C3 (NP_001008743). Peptide sequence PTSIMDHSISPFMRKGMPGDWKNYFTVAQNEEFDKDYQKKMAGSTLTFRT | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
442038 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title