missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric Acid, 50% Weight Solution, Chem-Lab Product Code.: 11959252
compliance-icons

Sulfuric Acid, 50% Weight Solution, Chem-Lab

Product Code. 11959252
1 L, Polyethylene, 1 L
Click to view available options
Quantity:
1 L
Packaging:
Polyethylene
Unit Size:
1 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11959252

Brand: Chem Lab CL02.2623.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Ideal for TMAR and as a solvent for caprolactam. Sulfuric Acid, 50% Weight Solution, Chem-Lab is a solution of 698g H2SO4 per liter of water for use as a reagent and neutralizing agent in laboratory applications.

Sulfuric acid is a highly corrosive, strong mineral acid that shows different properties depending upon its concentration. It is a pungent-ethereal, colorless to slightly yellow viscous liquid. Sulfuric acid is a diprotic acid that is hygroscopic. Concentrated sulfuric acid has strong dehydrating and oxidizing properties. Dilute sulfuric acid is a good electrolyte.

Specifications

Appearance Colorless
pH <1
CAS 7664-93-9, 7732-18-5
Composition 698g H2SO4 per 1L H2O
Melting Point 0°C
Boiling Point 119°C
Concentration 50% by weight
Density 1.39g/mL
Odor Odorless
Packaging Polyethylene
Quantity 1 L
Molecular Formula H2O4S
Physical Form Liquid
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Solubility Information Soluble in Water
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.08g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid 50 weight % solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric Acid, 50% Weight Solution, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.