Learn More
Sulfuric Acid, 4M, Chem-Lab Product Code.: 11923313
compliance-icons
Sulfuric Acid, 4M, Chem-Lab
Product Code. 11923313
1 L, 1 L
Click to view available options
Quantity:
1 L
10 L
Unit Size:
1 L
10 L
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11923313
Brand: Chem Lab CL05.2624.1000
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Ideal for titrations in laboratory applications. Sulfuric Acid, 4M, Chem-Lab is a solution of 392.32g H2SO4 per liter of water for use in volumetric analysis. Also known as battery acid, which is used in lead-acid batteries.
Sulfuric acid is a highly corrosive, strong mineral acid that shows different properties depending upon its concentration. It is a pungent-ethereal, colorless to slightly yellow viscous liquid. Sulfuric acid is a diprotic acid that is hygroscopic. Concentrated sulfuric acid has strong dehydrating and oxidizing properties. Dilute sulfuric acid is a good electrolyte.
Chemical Identifiers
CAS 7664-93-9, 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
Specifications
pH <1
Color Colorless
Composition 392.32g H2SO4 per 1L H2O
Melting Point 0°C
Boiling Point 100°C
Concentration 4mol/L
Density 1.23g/mL
Odor Odorless
Packaging Polyethylene
Normality 8N (±0.016, 20°C)
Quantity 1 L
Physical Form Liquid
Solubility Information Soluble in Water
Formula Weight 98.08g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid 4 mol/l
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid
Product Content Correction
Product Title
Sulfuric Acid, 4M, Chem-Lab >