Learn More
Sulfuric Acid, 0.05M, Reagecon™ Product Code.: 11583022
Sulfuric Acid, 0.05M, Reagecon™
BIOPHARMA
Product Code. 11583022 Shop All Reagecon Products
1 L, 1L
Click to view available options
Quantity:
1 L
Packaging:
1L
Product Code. 11583022
Brand: Reagecon SU20051
Specifications
Concentration 0.05M
Packaging 1L
Normality 0.1N
Quantity 1 L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberSU200524E1 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog NumberSU20051
Lot NumberSU200523K1 Certificate TypeCertificate of Analysis Date28/11/2023 Catalog NumberSU20051
Lot NumberSU200523G1 Certificate TypeCertificate of Analysis Date27/10/2023 Catalog NumberSU20051
Lot NumberSU200523C1 Certificate TypeCertificate of Analysis Date03/08/2023 Catalog NumberSU20051
Lot NumberSU200522K1 Certificate TypeCertificate of Analysis Date04/04/2023 Catalog NumberSU20051
Product Content Correction
Product Title
Sulfuric Acid, 0.05M, Reagecon™ >