Learn More
Sulfuric Acid, 0.01M, Chem Lab™
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
Specifications
Specifications
Appearance | Colorless |
pH | <2 |
Composition | Sulfuric acid 0.01mol/L |
Concentration | 0.981g H2SO4/l H2O = 0.02 N (±0.00004/20°C) |
Density | 1g/mL |
Odor | Odorless |
Quantity | 1 L |
Physical Form | Liquid |
Solubility Information | Soluble in Water |
Product Content Correction
Product Title
Sulfuric Acid, 0.01M, Chem Lab™ >