Learn More
Sulfur Certified Reference Material, UKAS ISO 17025 & ISO 17034 Certified, Sulfur in light mineral oil, 0.0010 % (10ppm), 100mL., Paragon Scientific™
Suitable for calibration, verification or for use in quality control procedures used for analysing sulfur content in petroleum products, with sulfur content ranging from 0 (Blank) through to 5.0000% (50000ppm) for each material.
Brand: Paragon Scientific LMSU-02
This item is not returnable.
View return policy
Description
- Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation.
- Highest level of accreditation guarantee, providing the most credible certified data available worldwide.
- Suitable for but not limited to internationally recognised test methods and protocol: ASTM D2622, ASTM D4294, ASTM D5453, ASTM D7039, IP 336, IP 496, IP 497.
- Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data.
- Supplied in 100 mL volume, tamper-evident glass packaging, providing assurance of sample integrity.
- Manufactured in the United Kingdom.
- 12 month shelf life.
Product Content Correction
Product Title
Nominal values quoted. Certified Values will vary from batch to batch.