missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUDS3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | SUDS3 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16844405
|
Proteintech
25845-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16834405
|
Proteintech
25845-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SDS3 is a subunit of the histone deacetylase.Specifications
SUDS3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9H7L9 | |
64426 | |
SUDS3 Fusion Protein Ag22961 | |
Primary | |
-20°C | |
SUDS3 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SAP45, SDS3, SUDS3 | |
SUDS3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title