missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic acid, Free acid, ≥99%, Ultrapure
CAS: 110-15-6 | C4H6O4 | 118.088 g/mol
900.00 NOK - 2145.00 NOK
Chemical Identifiers
CAS | 110-15-6 |
---|---|
Molecular Formula | C4H6O4 |
Molecular Weight (g/mol) | 118.088 |
MDL Number | MFCD0000278 |
InChI Key | KDYFGRWQOYBRFD-UHFFFAOYSA-N |
Synonym | succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid |
PubChem CID | 1110 |
ChEBI | CHEBI:15741 |
IUPAC Name | butanedioic acid |
SMILES | C(CC(=O)O)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
15875188
|
Thermo Scientific
J21900.36 |
500 g | Plastic bottle | N/A | ||||||
15855338
|
Thermo Scientific
J21900.A1 |
1 kg | Plastic bottle | N/A | ||||||
15865338
|
Thermo Scientific
J21900.A7 |
5 kg | Plastic bottle | N/A | ||||||
Description
Chemical Identifiers
110-15-6 | |
118.088 | |
KDYFGRWQOYBRFD-UHFFFAOYSA-N | |
1110 | |
butanedioic acid |
C4H6O4 | |
MFCD0000278 | |
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid | |
CHEBI:15741 | |
C(CC(=O)O)C(=O)O |
Specifications
110-15-6 | |
235°C | |
HOOCCH2CH2COOH | |
1754069 | |
KDYFGRWQOYBRFD-UHFFFAOYSA-N | |
butanedioic acid | |
1110 | |
118.09 | |
1.56 | |
Crystalline Powder | |
Succinic acid |
186°C to 188°C | |
C4H6O4 | |
MFCD0000278 | |
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid | |
C(CC(=O)O)C(=O)O | |
118.088 | |
CHEBI:15741 | |
≥99% | |
>110°C | |
1.56 |
Safety and Handling
P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
H318-H335
missing translation for 'einecsNumber' : 203-740-4
missing translation for 'rtecsNumber' : WM4900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Succinic acid, Free acid, ≥99%, Ultrapure