Description
Substance P is used as a neurokinin NK-1 receptor agonist, neurokinin undecapeptide neurotransmitter and neuromodulator, also used in cancer cell proliferation and that inhibition of this component can induce apoptosis in cancer cells and decrease the steady state of Her2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Purification Method | HPLC |
Format | Powder |
For Use With (Application) | A neurokinin NK-1 receptor agonist |
Molecular Weight (g/mol) | 1347.64 |
Product Type | Substance P |
Quantity | 25 mg |
Regulatory Status | RUO - research use only |
Sequence | Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2 |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
Q15L007 | Certificate of Analysis | 17/05/2025 | J63489.MB,J63489.#0 |
Q24L010 | Certificate of Analysis | 17/05/2025 | J63489.MB,J63489.#0 |
R13L021 | Certificate of Analysis | 17/05/2025 | J63489.MB,J63489.#0 |
R13L019 | Certificate of Analysis | 17/05/2025 | J63489.MB,J63489.#0 |
N12L009 | Certificate of Analysis | 14/02/2025 | J63489.MB,J63489.#0 |