Learn More
Memmert™ Subframe Adjustable In Height Up To 500 mm, For S450
Use these frames with Memmert convection ovens. Memmert™ Convection and Universal Oven Subframes are adjustable in height to fit your needs.
Brand:  Memmert™ S450B29753
Product Code. 16325847
Description
Description
  • Adjustable Subframe
  • Memmert Oven size 30 to 75: height 600mm
  • Size 110 to 450: height 500mm
Specifications
Specifications
Models U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, Model 450
22 in. (Size 110 to 160), 26 in. (Size 30 to 75)
560 mm (Size 110 to 160), 660 mm (Size 30 to 75)
Subframe
Product Title
Memmert™ Subframe Adjustable In Height Up To 500 mm, For S450 >