Learn More
Cole-Parmer™ Tube Holder
Tube Holder for Use with Microtitre Plate Shakers SSM5 and SSL5.
Brand:  Cole-Parmer™ 51900-99
 View more versions of this product
Product Code. 12693335
  • 1960.00 NOK / Each
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Description
  • Accessory tube holders accommodate 0.2 mL or 0.5 mL microcentrifuge tubes.
  • Two tube holders can be used at any one time and securely held in place by the non-slip mat.
  • Stuart™ Tube Holder; For Use with Microtitre Plate Shakers where samples require controlled temperature and/or humidity conditions, ideal for placement inside CO2incubators.
Specifications
Specifications
0.2 mLTubes
Tube Holder
1 Ea.
0.2 mL
Stuart™ Microtitre Plate Shakers SSM5 and SSL5
Product Title
Cole-Parmer™ Tube Holder > Tube Holder; For Use With: For 0.2 mL Tubes