Learn More
Cytiva Strip Holder
30440.00 NOK - 31505.00 NOK
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | ||||||
10690216
|
Cytiva
80-6416-30 |
11 cm |
31505.00 NOK
Pack of 6 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
||||||
10301325
|
Cytiva
80-6416-11 |
7 cm |
30440.00 NOK
Pack of 6 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
||||||
Description
Specifications
Strip Holder | |
1 Pack |
Product Title
Cytiva Strip Holder