Learn More
Invitrogen™ Streptavidin Protein, Rhodamine
Protein
Brand: Invitrogen™ 21724
2090.10 NOK valid until 2025-06-27
Use promo code "24112" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Streptavidin Protein, Rhodamine conjugate, 21724, from Invitrogen., 21724, from Invitrogen. Species Reactivity: ; Applications: ELISA, Immunohistochemistry, Western Blot.
Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Thermo Scientific Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.
Specifications
Rhodamine | |
PBS with no preservative | |
RUO | |
Lyophilized | |
purified | |
Streptavidin |
Tag | |
1 mg | |
4°C | |
ELISA, Immunohistochemistry, Western Blot | |
N/A |