missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Streptavidin - AP

Streptavidin - AP

Brand:  Molecular Probes™ 434322

2371.20 NOK valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 10573975

  • 3310.00 NOK / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Reactivity: AP-SA reacts with biotin.

Applications: Immunohistochemistry, Western blotting, and ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.5 mg/mL
3 mg
Wet Ice
Streptavidin Conjugate (fluorescent)
AP
Store in refrigerator (2–8°C).
Liquid
AP (Alkaline Phosphatase)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.