Learn More
Gosselin™ Straight Container, Hinged Cap
Corning™ Gosselin™ Straight Container, Hinged Cap is lightweight and break-resistant.
Brand:  Gosselin™ FL35-02
Product Code. 15482801
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Break-resistant polypropylene construction
  • Lightweight
  • Auto-stable hinged cap
  • Transparent, or blue
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
35 mL
Hinged Cap
950/Cs.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Gosselin™ Straight Container, Hinged Cap > 35mL