Learn More
Thermo Scientific™ Storerooms
Organize supplies, save valuable bench space using these six or nine drawer Storerooms. Order an empty cabinet or one fully stocked with vials and closures.
525.00 NOK - 7073.50 NOK
Product Code | Brand | Type | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Price | Quantity & Availability | |||||
12423578
|
Thermo Scientific™
C4000-MS |
Empty Mini-Storeroom Cabinet | Cabinet Only |
525.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
12440328
|
Thermo Scientific™
C4075-500 |
Empty Storeroom Cabinet | Cabinet Only | N/A | ||||||
12413788
|
Thermo Scientific™
C4011-688W |
Mini-Storeroom, Stocked | Vial: C4011-1W, Closure: C4011-1AP | N/A | ||||||
15270379
|
Thermo Scientific™
C5075-219 |
9 Drawer Storeroom, Stocked | Vial: C5000-1, Closure: C5000-54B | N/A | ||||||
Description
Organize your supplies and save valuable bench space with Thermo Scientific™ National™ 8mm Standard Opening Screw Thread Vial Storerooms. Storerooms are available in six and nine drawer models. The six drawer mini-storeroom holds 500 vials and closures; the nine drawer full size storeroom holds 2000 vials and closures. Order an empty cabinet or one fully stocked with vials and closures.
Choose from a variety of configurations to meet your needs.Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.