Learn More
BRAND™ Storage/ Dispenser Box
Ideal for storing and dispensing small laboratory items
Brand:  BRAND™ 131902
2165.14 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Product Code. 10642822
  • 2325.00 NOK / Each
Everyday Lab Essentials
Use promo code 24132 at checkout
View Available Promotions
Only null left
Estimated Shipment: 28-07-2025
Description
  • Ideal for storing and dispensing small laboratory items like pipette tips, microcentrifuge tubes, Pasteur pipettes, etc.
  • Removable top cover with hinged front cover
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    White and Clear
    PMMA
    165 x 152 x 178 mm
    Certificates
    Promotions
    Promotions
    • Multiple Manufacturers
      From wipes to waste bins, labels to liners – save on them all.
      Promo Code: 24132
      Expires2025-08-24
      New Offer
      Shop Now
    Product Content Correction
    Product Title
    BRAND™ Storage/ Dispenser Box > Dimensions (L x W x H): 165 x 152 x 178 mm