Learn More
Thermo Scientific™ Storage Canes
GREENER_CHOICE
Store various cryostorage tubes in these storage canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
Brand:  Thermo Scientific™ 5015-0001
196.40 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 10100341
  • 297.00 NOK / Pack of 12
Get 3 for 2
Use promo code 24112 at checkout
View Available Promotions
Only null left
Estimated Shipment: 30-06-2025
Add to basket
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Description
Store Thermo Scientific™ Nalgene™ or Thermo Scientifc™ Nunc™ cryostorage tubes in Thermo Scientific™ Storage Canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
  • Able to store tubes protected with Thermo Scientific™ CryoFlex™ tubing
  • Compatible with color coders that attach to the end of the cane and can be written on for quick identification and inventory management
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Stepped
Cryo Storage Cane
11.31 in.
Aluminum
12/Pk., 48/Cs.
30 cm
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number7280254010 Certificate TypeCertificate of Compliance Date25/02/2025 Catalog Number5015-0001
Lot Number7282717010 Certificate TypeCertificate of Compliance Date25/02/2025 Catalog Number5015-0001
Lot Number7285388010 Certificate TypeCertificate of Compliance Date25/02/2025 Catalog Number5015-0001
Lot Number7293406010 Certificate TypeCertificate of Compliance Date25/02/2025 Catalog Number5015-0001
Lot Number7299118010 Certificate TypeCertificate of Compliance Date25/02/2025 Catalog Number5015-0001
Promotions
Promotions
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Thermo Scientific™ Storage Canes > CryoCane; Stepped; Aluminum; 290mm
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis