Learn More
ratiolab™ Stoppers for Test Tubes
Close your test tubes with these highly secure stoppers.
Brand: ratiolab™ 351490G
This item is not returnable.
View return policy
Description
- Provides very tight seal due to two sealing rings
Specifications
11 to 12 mm | |
Green | |
1000 Stoppers/Bag |
Polyethylene | |
Bag |
Product Content Correction
Product Title
ratiolab™ Stoppers for Test Tubes > Stoppers for Test Tubes; Color: Green; Diameter (Metric) Outer: 11 to 12 mm