Learn More
Poulten Graf™ Stopper Caps
Brand:  Poulten Graf™ 101 08917K
 View more versions of this product
Product Code. 10583622
  • 486.00 NOK / Pack of 10
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Specifications
Stopper Cap
5 mL
1 Ea.
Documents
One moment while we fetch your results.
Product Title
Poulten Graf™ Stopper Caps > Capacity: 5mL