Learn More
Stopcock Assembly and Cover for Nalgene™ Transparent Polycarbonate Classic Design Desiccator
Get replacement parts for the Nalgene™ Transparent Polycarbonate Classic Design Desiccator
Brand: Thermo Scientific Nalgene 715311-0250
4784.80 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Get the Stopcock Assembly and Cover replacement parts for the Nalgene™ Transparent Polycarbonate Classic Design Desiccator, Cat #5311-0250. The Nalgene™ Transparent Polycarbonate Classic Design Desiccator maintains an airtight seal without the need of vacuum grease and can be used under full-vacuum and non-vacuum conditions, and is an excellent alternative to glass.
Description:
- Removable or replaceable stopcock
- Port capped if removed (cap included)
Specifications
Replacement Part | |
Cover for port on dessicator, polycarbonate |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1404518 | Certificate of Compliance | 07/04/2024 | 715311-0250 |
1394455 | Certificate of Compliance | 16/12/2023 | 715311-0250 |
1371109 | Certificate of Compliance | 29/09/2022 | 715311-0250 |
1347495 | Certificate of Compliance | 04/02/2022 | 715311-0250 |
1313101 | Certificate of Compliance | 04/03/2021 | 715311-0250 |