Learn More
Lenz Laborglasintrumente Stirrer with Stirrer Bearing
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 06310502
Product Code. 10142643
  • 180.20 NOK / Each
Estimated Shipment: 13-11-2024
to see stock.
Explore more special offers
Description
  • Shaft with blade and PTFE security screw
  • Gastight closure of vacuum stirring apparatuses up to 10-3 mbar at a continuous operation speed of max.
  • 800 rpm, for round bottom flasks 250-2000 mL
  • Shaft made of DURAN™ tubing
Specifications
Specifications
Blade
For round bottom flasks 250 to 2000 mL
PTFE