Learn More
Merck Stirred Cell Amicon
Brand:  Merck 5112
Product Code. 10219001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
60mL
1 Each
Stirred Cell Amicon
2 L
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Merck Stirred Cell Amicon > Capacity: 2L