Learn More
STIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15218-1-AP-150UL
 View more versions of this product
Product Code. 16884304
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
STIP1 is an adaptor protein that coordinates the functions of HSP70 and HSP90 in protein folding. It is thought to assist in the transfer of proteins from HSP70 to HSP90 by binding both HSP90 and substrate-bound HSP70. STIP1 also stimulates the ATPase activity of HSP70 and inhibits the ATPase activity of HSP90, suggesting that it regulates both the conformations and ATPase cycles of these chaperones (Song and Masison, 2005 ).
Specifications
Specifications
STIP1
Polyclonal
Unconjugated
STIP1
HOP, Hsc70/Hsp90 organizing protein, IEF SSP 3521, P60, STI1, STI1L, STIP1
Rabbit
Antigen Affinity Chromatography
RUO
10963, 192277, 20867
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35814, P31948, Q60864
Stip1
STIP1 Fusion Protein Ag7388
150 μL
Primary
Human, Mouse, Rat