Learn More
Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™
BIOPHARMA
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
Brand:  Sampling Systems™ 8045R-1000S
 View more versions of this product
Product Code. 17478873
  • 2265.00 NOK / Pack of 25
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged in ultra-strong packing with easy to open design
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Large variety of different sizes available
  • Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
  • Positioned the ergonomic handle high up on the scoop
  • No Cross Contamination
  • No Cleaning required
Specifications
Specifications
Disposable Scoop
Individually Bagged
Single-use
Red
Polystyrene
1000 mL
Sterile
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number120333 Certificate TypeCertificate of Compliance Date27/02/2025 Catalog Number8045R-1000S
Lot Number119274 Certificate TypeCertificate of Compliance Date15/01/2025 Catalog Number8045R-1000S
Lot Number119725 Certificate TypeCertificate of Compliance Date14/10/2024 Catalog Number8045R-1000S
Lot Number119275 Certificate TypeCertificate of Compliance Date09/10/2024 Catalog Number8045R-1000S
Lot Number119273 Certificate TypeCertificate of Compliance Date19/08/2024 Catalog Number8045R-1000S
Product Title
Sampling Systems™ SteriWare™ Red Disposable PharmaScoop™ > Capacity, Metric: 1000mL