Learn More
Sampling Systems™ SteriWare™ PP White Disposable PharmaScoop™
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ PP White Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
Brand: Sampling Systems™ 8045P-1000
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged in ultra-strong packing with easy to open design
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Large variety of different sizes available
- Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
- Positioned the ergonomic handle high up on the scoop
- No Cross Contamination
- No Cleaning required
Specifications
Disposable Scoop | |
Individually Bagged | |
Single-use | |
White |
Polypropylene | |
1000 mL | |
Non-sterile |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
122325 | Certificate of Compliance | 05/06/2025 | 8045P-1000 |
120298 | Certificate of Compliance | 11/02/2025 | 8045P-1000 |
120296 | Certificate of Compliance | 27/01/2025 | 8045P-1000 |
120296 | Certificate of Compliance | 21/11/2024 | 8045P-1000 |
118549 | Certificate of Compliance | 18/11/2024 | 8045P-1000 |