Learn More
Sampling Systems™ SteriWare™ Disposable Tweezers
Pick up both small and large items with a special dual action design. Sampling Systems™ SteriWare™ Disposable Tweezers are ideal for picking up items up to 25mm wide.
Brand:  Sampling Systems™ 8025H-115
 View more versions of this product
Product Code. 15839421
  • 769.00 NOK / Pack of 100
Only null left
Estimated Shipment: 14-08-2025
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No Cross Contamination
  • No Cleaning required
Specifications
Specifications
HDPE
115 mm
Non-Sterile Forceps
Non-sterile
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number122907 Certificate TypeCertificate of Compliance Date25/06/2025 Catalog Number8025H-115
Lot Number120951 Certificate TypeCertificate of Compliance Date05/12/2024 Catalog Number8025H-115
Lot Number103341 Certificate TypeCertificate of Compliance Date10/06/2022 Catalog Number8025H-115
Lot Number069470 Certificate TypeDeclaration of Conformity Date19/01/2023 Catalog Number8025H-115
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Disposable Tweezers > Product Type: Non-Sterile Forceps