Learn More
Sampling Systems™ SteriWare™ Disposable Powder Sampler
BIOPHARMA
Capable of being used straight out of the bag. Sampling Systems™ SteriWare™ Disposable Powder Sampler is ideal for taking large volume samples from powders or granules.
Brand:  Sampling Systems™ 8240H-1000
Product Code. 15819511
  • 3505.00 NOK / Pack of 20
Only null left
Estimated Shipment: 04-08-2025
Description
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Suitable for use in ATEX areas
  • Push the sampler into the product, rnsuring the sampler is in the closed position
  • Rotate handle to open up the slot
  • After leaving the sampler for several seconds to allow powder to enter, rotate handle to close the sampler
Specifications
Specifications
HDPE
100 mL
40 mm
1000 mm
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number118162 Certificate TypeCertificate of Compliance Date19/06/2024 Catalog Number8240H-1000
Lot Number55199 Certificate TypeCertificate of Compliance Date30/09/2022 Catalog Number8240H-1000
Lot Number067515 Certificate TypeCertificate of Compliance Date16/08/2022 Catalog Number8240H-1000
Lot Number068257 Certificate TypeDeclaration of Conformity Date05/12/2022 Catalog Number8240H-1000
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Disposable Powder Sampler > Capacity: 100mL