Learn More
Sampling Systems™ SteriWare™ Disposable Liquid Funnel
Ideal for laboratories, pharmaceutical environments, cleanrooms and chemical manufactruing. Sampling Systems™ SteriWare™ Disposable Liquid Funnel comes individually bagged after being moulded and packed in a Class 100,000 Cleanroom.
Brand:  Sampling Systems™ 8370H-100S
 View more versions of this product
Product Code. 15360044
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Constructed of HDPE
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Specifications
100 mm
12 mm
Sterile
Liquid Funnel
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number121370 Certificate TypeCertificate of Compliance Date11/03/2025 Catalog Number8370H-100S
Lot Number118487 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number8370H-100S
Lot Number117341 Certificate TypeCertificate of Compliance Date25/01/2024 Catalog Number8370H-100S
Lot Number067688 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8370H-100S
Lot Number117867 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number8370H-100S
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Disposable Liquid Funnel > ItemDescription: Yes