Learn More
Sampling Systems™ SteriWare™ Blue Disposable PharmaScoop™
BIOPHARMA
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Blue Disposable PharmaScoop™ is ideal for both sampling and dispensing powders and granules.
Brand:  Sampling Systems™ 8045B-60
 View more versions of this product
Product Code. 15859431
  • 1525.00 NOK / Pack of 100
Only null left
Estimated Shipment: 29-08-2025
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged in ultra-strong packing with easy to open design
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Large variety of different sizes available
  • Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
  • Positioned the ergonomic handle high up on the scoop
  • No Cross Contamination
  • No Cleaning required
Specifications
Specifications
Disposable Scoop
Individually Bagged
Single-use
Blue
Polypropylene
60 mL
Non-sterile
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number122053 Certificate TypeCertificate of Compliance Date26/03/2025 Catalog Number8045B-60
Lot Number121313 Certificate TypeCertificate of Compliance Date14/03/2025 Catalog Number8045B-60
Lot Number107994 Certificate TypeCertificate of Compliance Date28/02/2023 Catalog Number8045B-60
Lot Number102618 Certificate TypeCertificate of Compliance Date16/08/2022 Catalog Number8045B-60
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Blue Disposable PharmaScoop™ > Capacity, Metric: 60mL